missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ DexPure Vinyl Gloves

Product Code. 12626093
Click to view available options
Size:
Small
Medium
Large
X-Large
Powdered:
Powdered
Color:
Blue
Translucent
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 12626093

Brand: Honeywell Safety Products™ 4580001XL

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Convenient and disposable, these translucent vinyl gloves can be used for a variety of tasks from the laboratory to the kitchen. Honeywell Safety Products™ DexPure Vinyl Gloves also have a smooth finish that provides great dexterity.

  • Translucent vinyl glove.
  • Smooth finish for great dexterity.
  • Certified for food contact and treated to prevent bacteria.
  • Suitable for the following:
    • Product inspection.
    • Precision assembly.
    • Laboratory work.
    • Handling of paints, inks and dyes.
  • Single use.
  • AQL:1.5.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE CATEGORY II Intermediary risks, EN 420-2003 EEC Basic standards, EN 374-2003 Chemical hazards, EN 455-1&2 Medical Standards, European directive n°1934/2004 : Food contact, EC Category PPE 3, EC Certificate Number 0075/797/162/11/08/0465, EC Attestation Number 0075/797/162/11/08/0465
Classification Dexterity: 5
Description AQL:1.5
Color Translucent
Disposable Single-use
Material Vinyl
Powdered Powdered
Product Type Gloves
Size X-Large
Thickness (Metric) 0.1 mm
For Use With (Application) Automotive, Aviation, Catering, Chemical, Food, Industrial Cleaning, Laboratory, Maintenance, Medical, Pharmaceutical, Petrochemical, Printing and Water Treatment industries.
Quantity 100/PK
Length (Metric) 25 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.