Learn More
Dexamethasone Phosphate, TRC
Product Code. 18073840
Change view
Click to view available options
Quantity:
25 mg
100 mg
500 mg
Unit Size:
100mg
25mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18073840 100 mg 100mg
18011450 25 mg 25mg
18022450 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18073840 Supplier Toronto Research Chemicals Supplier No. TRCD298835100MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 312-93-6
Molecular Formula C22 H30 F O8 P
Molecular Weight (g/mol) 472.44
Synonym Dexamethasone Phosphate, Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,17,21-trihydroxy-16α-methyl-, 21-(dihydrogen phosphate), Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,17,21-trihydroxy-16α-methyl-, 21-phosphate, Betnelan phosphate, Dexamethasone 21-dihydrogen phosphate, Dexamethasone 21-phosphate, Dexamethasone orthophosphate, Micromethason, Wymesone
IUPAC Name [2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] dihydrogen phosphate
SMILES C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COP(=O)(O)O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C22H30FO8P/c1-12-8-16-15-5-4-13-9-14(24)6-7-19(13,2)21(15,23)17(25)10-20(16,3)22(12,27)18(26)11-31-32(28,29)30/h6-7,9,12,15-17,25,27H,4-5,8,10-11H2,1-3H3,(H2,28,29,30)/t12-,15+,16+,17+,19+,20+,21+,22+/m1/s1
Formula Weight 472.1662
Percent Purity >95%
Chemical Name or Material Dexamethasone Phosphate
Grade HPLC