Learn More
Dexamethasone Acid, TRC
Product Code. 18002050
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18002050 10 mg 10mg
17939759 50 mg 50mg
18050800 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18002050 Supplier Toronto Research Chemicals Supplier No. TRCD29880510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 37927-01-8
Molecular Formula C21 H27 F O5
Molecular Weight (g/mol) 378.43
Synonym (11β,16α,17α)-9-Fluoro-11,17-dihydroxy-16-methyl-3-oxo-androsta-1,4-diene-17-carboxylic Acid, (-)-Dexamethasone acid, 9-Fluoro-11β,17-dihydroxy-16α-methyl-3-oxoandrosta-1,4-diene-17β-carboxylic acid, Dexamethasone Sodium Phosphate Imp. G (EP)
IUPAC Name (8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthrene-17-carboxylic acid
SMILES C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)O
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C21H27FO5/c1-11-8-15-14-5-4-12-9-13(23)6-7-18(12,2)20(14,22)16(24)10-19(15,3)21(11,27)17(25)26/h6-7,9,11,14-16,24,27H,4-5,8,10H2,1-3H3,(H,25,26)/t11-,14+,15+,16+,18+,19+,20+,21+/m1/s1
Formula Weight 378.1843
Percent Purity >95%
Chemical Name or Material (-)-Dexamethasone Acid
Grade HPLC