Learn More
Epredia™ Dewax and HIER Buffer L,M,H
Product Code. 15220715 Shop All Epredia Products
Change view
Click to view available options
Solution Type:
Buffer - High
Buffer - Low
Buffer - Medium
Unit Size:
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Solution Type Unit Size
15220715 Buffer - Medium 1 L
15200715 Buffer - High 1 L
15210715 Buffer - Low 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15220715 Supplier Epredia™ Supplier No. TA999DHBM
 Restricted Item
Estimated Shipment: 03-11-2026
Epredia™ Dewax and Heat Induced Epitope Retrieval (HIER) Buffers simultaneously automate deparaffinization, rehydration and epitope retrieval in the Epredia™ PT Module.
Automated deparaffinization (dewaxing) and rehydration of IHC slides is an ideal way to decrease turn-around time and improve lean workflow. Aqueous deparaffinization is the method of choice for IHC instruments today but often yields incomplete deparaffinization. The Dewax and Heat Induced Epitope Retrieval (HIER) Buffers simultaneously automate deparaffinization, rehydration and epitope retrieval in the Epredia PT Module. The new proprietary formulation eliminates manual steps and improves turnaround time. In addition the results are concordant to traditional methods meaning they can be quickly integrated into routine workflow.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Dewax and HIER Buffer
Quantity 10 x 100 mL
Solution Type Buffer - Medium
compliance-icons compliance-icons
Product Identifier
  • DEWAX AND HIER BUFFER M (15X CONCENTRATE) 10X100ML
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H319-Causes serious eye irritation.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • EUH208-Contains 2-methyl-3-isothiazolone. May produce an allergic reaction.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Tetrahydrofurfuryl alcohol, EDTA, Tetrasodium EDTA, 2-Methyl-3-isothiazolone