Learn More
Day Impex™ Glass Dewar Refill
Product Code. 11867442
Change view
Click to view available options
Capacity (Metric):
170 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11867442 170 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11867442 Supplier Day Impex™ Supplier No. 210014
  • ONLINE EXCLUSIVE PRICE
    189.57€
    145.39€ / Pack of 1
    Save 44.18€
    23% Off
Estimated Shipment: 04-02-2027
Designed with practicality and safety in mind. Day Impex™ Glass Dewar Refill is ultra-low evacuated to allow for safe and efficient handling of all liquified gases, freezing mixtures, and specimens.
  • Borosilicate glass inner vessels
  • Ultra-low evacuated
  • Safe and efficient handling of all liquified gases, freezing-mixtures and specimens
Specifications
Depth (Metric) Interior 50 mm
Capacity (Metric) 170 mL
Diameter (Metric) Inner 77 mm
Height (Metric) Inner 50mm