Learn More
Thermo Scientific™ DeviceLink™ HUB Connectivity Kit for Thermo Scientific CO2 Incubators
Connect your lab equipment and experience the Thermo Fisher Cloud™solutions
Alert:
Available in NA, EU and EFTA countries only. Please check with your local sales representative for regional availability.
Description
Description
Connectivity is enhancing workflows and increasing efficiency and effectiveness within the lab.   Thermo Scientific™ DeviceLink™ Hub, our easy-to-install laboratory solution, works with the Thermo Fisher Cloud to monitor conditions.   For CO2 incubators, the DeviceLink™ Hub monitoring solution reports chamber temperature, gas levels, alarms, and alerts from the incubator data output connection.
Key features and benefits
  • View critical lab equipment data and equipment notifications on you mobile and web applications.
    • Chamber temperature
    • Gas levels
    • Alarm and alerts notification
  • Access data anytime, anywhere—data and alarms from monitored lab equipment are securely stored on your Thermo Fisher Cloud account and sent to the user via email and mobile-phone notification
  • InstrumentConnect™ streamlined dashboard-style reporting reduces time spent interpreting data
  • Unlike traditional methods, the DeviceLink Hub monitoring solution operates on a client-cloud architecture, requiring minimal IT support
  • No need to install sensors—the only plug-in is the equipment data port to read the parameters directly from the factory-installed sensors
Security
Security is our highest priority. We understand that the security of data and protocols are critical, both to your research and to the publication process. To help ensure high levels of protection and security, we’ve built Thermo Fisher Cloud on the world-class Amazon Web Services™ (AWS) cloud-computing platform. With AWS, all of your data is stored in highly secure AWS data centers managed by compliance programs and audit safeguards.
Monitored Data
Telemetry
  • Chamber temperature (oC)
  • CO2 levels (%)
  • Optional O2 levels (%) 
Alarms & events
  • High/low alarms
  • Door open
  • Low water (with models with humidity reservoir water level sensor)
  • Sterilization Cycle
  • AutoStart
  • Other system notifications
Compatibility
  • Thermo Scientific Heracell™ 150i and 240i CO2 incubators,
  • Thermo Scientific Heracell VIOS 160i and 250i CO2 incubators, Forma™ i160 and i250 CO2 incubators, Forma Series 3 CO2 incubators, All models with connection through the standard USB output
Setup
Follow a simple three-step installation and configuration to your internal WI-FI account
  1. Plug in and power on the device
  2. Connect the DeviceLink Hub hardware
  3. Configure the DeviceLink Hub solution through the simple-to-use mobile or desktop web interface
What is included?
  • (1) DeviceLinkHUB™ Remote Monitoring Module
  • (1) High Performance Data Cable
*Note: This kit is for use with Thermo Scientific CO2 Incubator units equipped iCAN™ touchscreen with RS232 or USB Data Port