Learn More
Fisherbrand™ Deuterium Lamps
Product Code. 15364078 Shop All Fisherbrand Products
Click to view available options
For Use With (Equipment):
ABI 757 759 783A 785A 1000S
Agilent 1100 DAD, 8453
Agilent 1100 VWD long life
Agilent 1100, 1200 DAD
Agilent 1100, 1200 G1315/G1365
Agilent 1290 DAD
Dionex PDA-100, PDA-3000, AD25
Dionex Ultimate UVD 3000
Hitachi L & U Series
Jasco UV970 975 1570 1575
Jasco V 500 530 550 560 570
PE Lambda 2 to 45 800 900 Bio
Shimadzu LC2010, D2
Shimadzu SPD10A, AVP, AV, AVVP
Shimadzu UV Spec,BioSpec-mini
Varian Prostar 325 335
Varian Prostar 330 DAD
Varian Prostar 340 345
Varian UV50/100/200 9050
Waters 2487 long life
Waters 996, 2996 PDA long Life
Waters Acquity 2489/2998
Unit Size:
Each
Product Code. 15364078
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Designed for compatibility with a variety of instruments. Fisherbrand™ Deuterium Lamps are individually tested, certified, and guaranteed to the same specifications as OEM.
  • Produced in state of the art manufacturing facilities approved by lamp manufacturers
  • Full traceability of manufactured products
  • Lamps available for all major instruments
Specifications
Light Source UV-A, UV-C
Wavelength 400/185 nm
Aperture Size (Metric) 0.5 mm
For Use With (Equipment) Waters 996, 2996 PDA long Life
Packaging Quantity 1 Ea.
Product Type Deuterium Lamp