Learn More
Deuterated Mix, 3-2135 in Toluene, Dr. Ehrenstorfer
Click to view available options
Quantity:
5x1 mL
Unit Size:
5mL
Specifications
Specifications
| Chemical Name or Material | 1-AminonaphthaleneD7∣2-Naphthylamine-d7∣4-Aminobiphenyl D9 |
| CAS | 78832-53-8, 93951-94-1, 344298-96-0 |
| Solvent or Matrix | Toluene |
| Quantity | 5x1 mL |
| Concentration or Composition (by Analyte or Components) | 4-Aminobiphenyl D9 25 μg/mL∣1-AminonaphthaleneD7 50 μg/mL∣2-Naphthylamine-d7 50 μg/mL |