missing translation for 'onlineSavingsMsg'
Learn More

behr Labor-Technik™ DET-Total Cyanide Application Kit

Product Code. 10700851
Click to view available options
Width (Metric):
1000 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10700851

Brand: behr Labor-Technik™ B00218115

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Width (Metric) 1000 mm
Height (Metric) 500mm
For Use With (Application) For Total Cyanide
Type DET-Total Cyanide Application Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.