Learn More
Desulfo Glucoraphanin, TRC
Specifications
Specifications
| Chemical Name or Material | Desulfo Glucoraphanin |
| CAS | 287966-62-5 |
| Molecular Formula | C12 H23 N O7 S2 |
| Quantity | 2.5 mg |
| Synonym | 1-Thio-1-[N-hydroxy-5-(methylsulfinyl)pentanimidate] β-D-Glucopyranose, 4-Methylsulfinylbutyldesulfoglucosinolate |
| InChI Formula | InChI=1S/C12H23NO7S2/c1-22(19)5-3-2-4-8(13-18)21-12-11(17)10(16)9(15)7(6-14)20-12/h7,9-12,14-18H,2-6H2,1H3/b13-8+/t7-,9-,10+,11-,12+,22?/m1/s1 |
| SMILES | CS(=O)CCCC\C(=N/O)\S[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O |
| IUPAC Name | [(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydropyran-2-yl] (1E)-N-hydroxy-5-methylsulfinyl-pentanimidothioate |
| Molecular Weight (g/mol) | 357.44 |
| Formula Weight | 357.0916 |