Learn More
Desmosterol, TRC
Product Code. 30011003
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011003 100 mg 100mg
30008671 25 mg 25mg
30003337 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011003 Supplier Toronto Research Chemicals Supplier No. TRCD296860100MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 313-04-2
Molecular Formula C27 H44 O
Molecular Weight (g/mol) 384.64
Synonym Cholesta-5,24-dien-3-ol, (3β)-, Cholesta-5,24-dien-3β-ol (8CI), (3β)-Cholesta-5,24-dien-3-ol, 24,25-Dehydrocholesterol, 24-Dehydrocholesterol, Desmosterol, NSC 226126, OR 942 (Sterol), Cholesta-5,24-dien-3-ol
IUPAC Name (3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylhept-5-en-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES C[C@H](CCC=C(C)C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C27H44O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h7,9,19,21-25,28H,6,8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1
Formula Weight 384.3392
Percent Purity >95%
Chemical Name or Material Desmosterol
Grade HPLC