missing translation for 'onlineSavingsMsg'
Learn More

Desmethylene Tadalafil, TRC

Product Code. 17951165
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17951165 5 mg 5mg
17953185 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17951165 Supplier Toronto Research Chemicals Supplier No. TRCD2919905MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 171489-03-5
Molecular Formula C21 H19 N3 O4
Molecular Weight (g/mol) 377.39
Synonym Pyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione, 6-(3,4-dihydroxyphenyl)-2,3,6,7,12,12a-hexahydro-2-methyl-, (6R,12aR)-, Pyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione, 6-(3,4-dihydroxyphenyl)-2,3,6,7,12,12a-hexahydro-2-methyl-, (6R-trans)-, (6R,12aR)-6-(3,4-Dihydroxyphenyl)-2,3,6,7,12,12a-hexahydro-2-methylpyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione, (6R,trans)-6-(3,4-dihydroxyphenyl)-2,3,6,7,12,12a-hexahydro-2-methyl-pyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione
IUPAC Name (2R,8R)-2-(3,4-dihydroxyphenyl)-6-methyl-3,6,17-triazatetracyclo[8.7.0.0^{3,8}.0^{11,16}]heptadeca-1(10),11,13,15-tetraene-4,7-dione
SMILES CN1CC(=O)N2[C@H](Cc3c([nH]c4ccccc34)[C@H]2c5ccc(O)c(O)c5)C1=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C21H19N3O4/c1-23-10-18(27)24-15(21(23)28)9-13-12-4-2-3-5-14(12)22-19(13)20(24)11-6-7-16(25)17(26)8-11/h2-8,15,20,22,25-26H,9-10H2,1H3/t15-,20-/m1/s1
Formula Weight 377.1376
Chemical Name or Material Desmethylene Tadalafil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.