missing translation for 'onlineSavingsMsg'
Learn More

Desmethoxy Yangonin, TRC

Product Code. 30354686
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354686 25 mg 25mg
30350738 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30354686 Supplier Toronto Research Chemicals Supplier No. TRCD29067025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15345-89-8
Molecular Formula C14 H12 O3
Molecular Weight (g/mol) 228.24
Synonym 2H-Pyran-2-one, 4-methoxy-6-[(1E)-2-phenylethenyl]-, 2H-Pyran-2-one, 4-methoxy-6-(2-phenylethenyl)-, (E)-, 2H-Pyran-2-one, 4-methoxy-6-styryl-, (E)- (8CI), Yangonin, demethoxy- (6CI), 4-Methoxy-6-[(1E)-2-phenylethenyl]-2H-pyran-2-one, 5,6-Dehydrokavain, 5,6-Dehydrokawain, Demethoxyyangonin, Desmethoxyyangonin, NSC 112161, NSC 68686, trans-5,6-Dehydrokawain
IUPAC Name 4-methoxy-6-[(E)-2-phenylethenyl]pyran-2-one
SMILES COC1=CC(=O)OC(=C1)\C=C\c2ccccc2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C14H12O3/c1-16-13-9-12(17-14(15)10-13)8-7-11-5-3-2-4-6-11/h2-10H,1H3/b8-7+
Formula Weight 228.0786
Chemical Name or Material Desmethoxyyangonin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.