missing translation for 'onlineSavingsMsg'
Learn More

Deschloroketamine Hydrochloride, TRC

Product Code. 30364072
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30364072 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30364072 Supplier Toronto Research Chemicals Supplier No. TRCD28958810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Deschloroketamine hydrochloride
CAS 4631-27-0
Molecular Formula C13 H17 N O . Cl H
Quantity 10 mg
Synonym Cyclohexanone, 2-(methylamino)-2-phenyl-, hydrochloride (1:1), Cyclohexanone, 2-(methylamino)-2-phenyl-, hydrochloride (7CI,8CI,9CI), 2'-Oxo-PCM hydrochloride, Deschloroketamine hydrochloride, 2-(Methylamino)-2-phenylcyclohexan-1-one hydrochloride, 2-(Methylamino)-2-phenyl-cyclohexanone hydrochloride
InChI Formula InChI=1S/C13H17NO.ClH/c1-14-13(10-6-5-9-12(13)15)11-7-3-2-4-8-11;/h2-4,7-8,14H,5-6,9-10H2,1H3;1H
SMILES Cl.CNC1(CCCCC1=O)c2ccccc2
IUPAC Name 2-(methylamino)-2-phenylcyclohexan-1-one;hydrochloride
Molecular Weight (g/mol) 239.74
Formula Weight 239.1077
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.