missing translation for 'onlineSavingsMsg'
Learn More

Deschloroethyl Bendamustine Hydrochloride, TRC

Product Code. 30135703
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135703 25 mg 25mg
30005453 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135703 Supplier Toronto Research Chemicals Supplier No. TRCD28836525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1797881-48-1
Molecular Formula C14 H18 Cl N3 O2 . Cl H
Molecular Weight (g/mol) 332.23
Synonym 1H-Benzimidazole-2-butanoic acid, 5-[(2-chloroethyl)amino]-1-methyl-, hydrochloride (1:1), Bendamustine Related Compound D (USP) as HCl, 4-[5-[(2-Chloroethyl)amino]-1-methyl-1H-benzimidazol-2-yl]butanoic acid hydrochloride
IUPAC Name 4-[5-(2-chloroethylamino)-1-methylbenzimidazol-2-yl]butanoic acid;hydrochloride
SMILES Cl.Cn1c(CCCC(=O)O)nc2cc(NCCCl)ccc12

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C14H18ClN3O2.ClH/c1-18-12-6-5-10(16-8-7-15)9-11(12)17-13(18)3-2-4-14(19)20;/h5-6,9,16H,2-4,7-8H2,1H3,(H,19,20);1H
Formula Weight 331.0854
Chemical Name or Material Deschloroethyl Bendamustine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.