Learn More
Desbutyl Bumetanide, TRC
Product Code. 17981345
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17981345 100 mg 100mg
17932065 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17981345 Supplier Toronto Research Chemicals Supplier No. TRCD288780100MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 28328-54-3
Molecular Formula C13 H12 N2 O5 S
Molecular Weight (g/mol) 308.31
Synonym Benzoic acid, 3-amino-5-(aminosulfonyl)-4-phenoxy-, Benzoic acid, 3-amino-4-phenoxy-5-sulfamoyl- (8CI), 3-Amino-4-phenoxy-5-sulfamoylbenzoic acid, Desbutylbumetanide, PF 1578
IUPAC Name 3-amino-4-phenoxy-5-sulfamoylbenzoic acid
SMILES Nc1cc(cc(c1Oc2ccccc2)S(=O)(=O)N)C(=O)O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C13H12N2O5S/c14-10-6-8(13(16)17)7-11(21(15,18)19)12(10)20-9-4-2-1-3-5-9/h1-7H,14H2,(H,16,17)(H2,15,18,19)
Formula Weight 308.0467
Chemical Name or Material Desbutyl Bumetanide