Learn More
Tocris Bioscience™ des-His1-[Glu9)-Glucagon (1-29) amide
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
des-His1-[Glu9]-Glucagon (1-29) amide is a glucagon receptor antagonist (pA2 = 7.2 for inhibition of glucagon-induced adenylyl cyclase activation in rat liver membranes); displays no agonist activity. Enhances glucose-stimulated pancreatic insulin release in vitro. Blocks added glucagon-induced hyperglycemia in normal rabbits without affecting glycogenolysis in vivo. Also blocks endogenous glucagon-induced hyperglycemia in streptozocin diabetic rats.
Specifications
Specifications
| Purity | 95% |
| Content And Storage | Store at -20°C |
| Description | des-His1-[Glu9)-Glucagon (1-29) amide |
| Molecular Weight (g/mol) | 3358.68 |
| Product Type | des-His1-[Glu9]-Glucagon (1-29) amide |
| Quantity | 1 mg |
| Sequence | Ser-Gln-Gly-Thr-Phe-Thr-Ser-Glu-Tyr-Ser-Lys-Tyr-Leu-Asp-Ser-Arg-Arg-Ala-Gln-Asp-Phe-Val-Gln-Trp-Leu-Met-Asn-Thr-NH2 |