Learn More
Deoxycholic Acid-24-13C, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Deoxycholic Acid (24-13 C) |
| CAS | 52886-37-0 |
| Molecular Formula | 13 C C23 H40 O4 |
| Quantity | 0.05 g |
| Synonym | Cholan-24-oic-24-13 C acid, 3,12-dihydroxy-, (3α,5β,12α)-, Deoxycholic-24-13 C acid, (3α,5β,12α)-3,12-Dihydroxycholan-24-oic-24-13 C acid, Deoxycholic acid-13 C, Deoxycholic-24-13 C acid, Deoxycholic acid (24-13 C), Deoxycholic acid (24-13 C) |
| InChI Formula | InChI=1 S/C24H40O4/c1-14(4-9-22(27)28)18-7-8-19-17-6-5-15-12-16(25)10-11-23(15,2)20(17)13-21(26)24(18,19)3/h14-21,25-26 H,4-13H2,1-3H3,(H,27,28)/t14-,15-,16-,17+,18-,19+,20+,21+,23+,24-/m1/s1/i22+1 |
| SMILES | C[C@H](CC[13 C](=O)O)[C@H]1 CC[C@H]2 [C@@H]3 CC[C@@H]4 C[C@H](O)CC[C@]4(C)[C@H]3 C[C@H](O)[C@]12 C |
| IUPAC Name | (4 R)-4-[(3 R,5 R,8 R,9 S,10 S,12 S,13 R,14 S,17 R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1 H-cyclopenta[a]phenanthren-17-yl](1 ^{13}C)pentanoic acid |
| Molecular Weight (g/mol) | 393.565 |
| Formula Weight | 393.296 g/mol |
| Show More |