Learn More
Munktell Filtrak™ Dental Napkins
Click to view available options
Dimensions (L x W):
145 x 230 mm
Unit Size:
Pack of 250
Description
- Creped grade 376 and coated on one side grade LabSorb Ultra
Specifications
Specifications
| Format | Sheet |
| Grade | LabSorb Ultra |
| Quantity | 200 Pk. |
| Type | Dental Napkin |
| Dimensions (L x W) | 145 x 230 mm |