Learn More
Smeg™ Demineralized Water Pump
Product Code. 12366023
Click to view available options
:
Demineralized Water Pump; For Use With (Equipment): Nonpressurized Water Tank; Product Type: Booster Pump
Product Code. 12366023
Brand: Smeg™ PAD
Only null left
Add to basket
Add to basket
Booster pump for non-pressurized, demineralized water.
Booster pump for non-pressurized, demineralized water.
  • Allows feeding machine with demineralized water drawn from a nonpressurized tank positioned at a minimum height of 85 cm from ground
Specifications
Product Type Booster Pump
For Use With (Equipment) Nonpressurized Water Tank