missing translation for 'onlineSavingsMsg'
Learn More

Demethylbleomycin A2 (Technical Grade), TRC

Product Code. 30372921
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30372921 1 mg
30377235 10 mg
30373780 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30372921 Supplier Toronto Research Chemicals Supplier No. TRCD5958151MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41089-03-6
Molecular Formula C54H81N17O21S3
Molecular Weight (g/mol) 1400.52
Synonym Bleomycin DM-A2, N1-[3-(Methylthio)propyl]bleomycinamide, (2R,3S,4S,5R,6R)-2-(((2R,3S,4R,5S,6S)-2-((1R,2S)-2-(6-Amino-2-((S)-3-amino-1-(((S)-2,3-diamino-3-oxopropyl)amino)-3-oxopropyl)-5-methylpyrimidine-4-carboxamido)-3-(((2R,3S,4S)-3-hydroxy-5-(((3R)-3-hydroxy-1-((2-(4-((3-(methylthio)propyl)carbamoyl)-[2,4'-bithiazol]-2'-yl)ethyl)amino)-1-oxobutan-2-yl)amino)-4-methyl-5-oxopentan-2-yl)amino)-1-(1H-imidazol-4-yl)-3-oxopropoxy)-4,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-4-yl Carbamate
IUPAC Name [(2R,5R)-2-[(2R,5S)-2-[(2S)-2-[[6-amino-2-[3-amino-1-[[(2S)-2,3-diamino-3-oxopropyl]amino]-3-oxopropyl]-5-methylpyrimidine-4-carbonyl]amino]-3-[[(2R,3S,4S)-3-hydroxy-5-[[(3R)-3-hydroxy-1-[2-[4-[4-(3-methylsulfanylpropylcarbamoyl)-1,3-thiazol-2-yl]-1,3-thiazol-2-yl]ethylamino]-1-oxobutan-2-yl]amino]-4-methyl-5-oxopentan-2-yl]amino]-1-(1H-imidazol-4-yl)-3-oxopropoxy]-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]oxy-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl] carbamate
SMILES CSCCCNC(=O)c1csc(n1)c2csc(CCNC(=O)C(NC(=O)[C@@H](C)[C@H](O)[C@@H](C)NC(=O)[C@@H](NC(=O)c3nc(nc(N)c3C)C(CC(=O)N)NC[C@H](N)C(=O)N)C(O[C@@H]4OC(CO)[C@@H](O)C(O)C4O[C@H]5OC(CO)[C@@H](O)C(OC(=O)N)C5O)c6c[nH]cn6)[C@@H](C)O)n2

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C54H81N17O21S3/c1-19-32(68-45(71-43(19)57)24(11-30(56)75)63-12-23(55)44(58)81)49(85)70-34(40(25-13-60-18-64-25)90-53-42(38(79)36(77)28(14-72)89-53)91-52-39(80)41(92-54(59)87)37(78)29(15-73)88-52)50(86)65-21(3)35(76)20(2)46(82)69-33(22(4)74)48(84)62-9-7-31-66-27(17-94-31)51-67-26(16-95-51)47(83)61-8-6-10-93-5/h13,16-18,20-24,28-29,33-42,52-53,63,72-74,76-80H,6-12,14-15,55H2,1-5H3,(H2,56,75)(H2,58,81)(H2,59,87)(H,60,64)(H,61,83)(H,62,84)(H,65,86)(H,69,82)(H,70,85)(H2,57,68,71)/t20-,21+,22+,23-,24?,28?,29?,33?,34-,35-,36+,37+,38?,39?,40?,41?,42?,52+,53-/m0/s1
Formula Weight 1399.4955
Chemical Name or Material Demethylbleomycin A2 (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.