Learn More
Delta4-Pregnene-3,20-diol, TRC
Product Code. 30412289
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30412289 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30412289 Supplier Toronto Research Chemicals Supplier No. TRCP7122601G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material delta4-Pregnene-3,20-diol
CAS 28507-76-8
Molecular Formula C21H34O2
Quantity 1 g
Synonym 3-Hydroxy-4-pregnen-20-ol, 3,20-Dihydroxypregn-4-ene
InChI Formula InChI=1S/C21H34O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h12-13,15-19,22-23H,4-11H2,1-3H3/t13-,15?,16-,17+,18-,19-,20-,21+/m0/s1
SMILES C[C@H](O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name (8S,9S,10R,13S,14S,17S)-17-[(1S)-1-hydroxyethyl]-10,13-dimethyl-2,3,6,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
Molecular Weight (g/mol) 318.49
Formula Weight 318.2559