missing translation for 'onlineSavingsMsg'
Learn More

Delta4-Pregnene-3,20-diol, TRC

Product Code. 30410066
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30410066 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410066 Supplier Toronto Research Chemicals Supplier No. TRCP712260100MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material delta4-Pregnene-3,20-diol
CAS 28507-76-8
Molecular Formula C21H34O2
Quantity 100 mg
Synonym 3-Hydroxy-4-pregnen-20-ol, 3,20-Dihydroxypregn-4-ene
InChI Formula InChI=1S/C21H34O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h12-13,15-19,22-23H,4-11H2,1-3H3/t13-,15?,16-,17+,18-,19-,20-,21+/m0/s1
SMILES C[C@H](O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name (8S,9S,10R,13S,14S,17S)-17-[(1S)-1-hydroxyethyl]-10,13-dimethyl-2,3,6,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
Molecular Weight (g/mol) 318.49
Formula Weight 318.2559
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.