missing translation for 'onlineSavingsMsg'
Learn More
Learn More
delta-Sarcoglycan Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2778-SP
This item is not returnable.
View return policy
Description
delta-Sarcoglycan Polyclonal specifically detects delta-Sarcoglycan in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| delta-Sarcoglycan | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 35 kDa dystrophin-associated glycoprotein, 35DAG, 35kD dystrophin-associated glycoprotein, CMD1L, CMD1LSG-delta, DAGD, delta-sarcoglycan, deltaSarcoglycan, delta-SG, dystrophin associated glycoprotein, delta sarcoglycan, LGMD2F, MGC22567, placental delta sarcoglycan, sarcoglycan, delta (35kD dystrophin-associated glycoprotein), sarcoglycan, delta (35kDa dystrophin-associated glycoprotein), SGCD, SGCDP, SGD | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6444 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Q92629 | |
| SGCD | |
| E. coli-derived recombinant human delta-Sarcoglycan isoform 1 Met1-Glu232 Accession # Q92629 | |
| 25 μg | |
| Primary | |
| Detects human delta-Sarcoglycan in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction