missing translation for 'onlineSavingsMsg'
Learn More

Delta-Guanido-Alpha-ketovaleric Acid, TRC

Product Code. 30396902
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396902 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396902 Supplier Toronto Research Chemicals Supplier No. TRCG41210010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material delta-Guanido-alpha-ketovaleric Acid
CAS 3715-10-4
Purity Grade Notes HPLC
Molecular Formula C6H11N3O3
Quantity 10 mg
Synonym 5-Guanidino-2-oxo-valeric Acid, 5-[(Aminoiminomethyl)amino]-2-oxopentanoic Acid, 2-Oxo-5-guanidinovaleric Acid, 5-Guanidino-2-oxovaleric Acid, alpha-Keto-delta-guanidinovaleric Acid, delta-Guanidino-alpha-oxovaleric Acid
InChI Formula InChI=1S/C6H11N3O3/c7-6(8)9-3-1-2-4(10)5(11)12/h1-3H2,(H,11,12)(H4,7,8,9)
SMILES NC(=N)NCCCC(=O)C(=O)O
IUPAC Name 5-carbamimidamido-2-oxopentanoic acid
Molecular Weight (g/mol) 173.17
Formula Weight 173.08
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.