Learn More
Delorazepam 4-Oxide, TRC
Specifications
Specifications
| Chemical Name or Material | 7-Chloro-5-(2-chlorophenyl)-1,3-dihydro-2H-1,4-benzodiazepin-2-one 4-Oxide |
| CAS | 2955-37-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C15 H10 Cl2 N2 O2 |
| Quantity | 10 mg |
| Synonym | 7-Chloro-5-(2-chlorophenyl)-1,3-dihydro-2H-1,4-benzodiazepin-2-one 4-Oxide, Lorazepam Imp. C (EP) |
| InChI Formula | InChI=1S/C15H10Cl2N2O2/c16-9-5-6-13-11(7-9)15(19(21)8-14(20)18-13)10-3-1-2-4-12(10)17/h1-7H,8H2,(H,18,20) |
| SMILES | [O-][N+]1=C(c2ccccc2Cl)c3cc(Cl)ccc3NC(=O)C1 |
| IUPAC Name | 7-chloro-5-(2-chlorophenyl)-4-oxido-1,3-dihydro-1,4-benzodiazepin-4-ium-2-one |
| Molecular Weight (g/mol) | 321.16 |
| Show More |