Learn More
Scotch-Brite™ Delicate Surface Cleaning Pads 98
Product Code. 17198820
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 60
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17198820 White Pack of 60
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17198820 Supplier Scotch-Brite™ Supplier No. 7100169976
  • ONLINE EXCLUSIVE PRICE
    246.00€
    129.14€ / Pack of 60
    Save 116.86€
    47% Off
View all promotions
Estimated Shipment: 19-11-2026
Light duty scouring pad designed for cleaning delicate surfaces. Soft, non-woven fibers and mild abrasive gently, but thoroughly, clean most surfaces without scratching including ceramic and glass.
  • White pad for delicate surface cleaning
  • Ultra soft, without abrasive particles
  • Easy to rinse/wash after use
  • Ideal for cleaning porcelain, bathtubs, washbasins, sinks, tile walls, and many more
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleaning Pad
Color White
Dimensions (L x W) 224 x 158 mm
For Use With (Application) Cleaning Delicate Surfaces
Length (Metric) 224 mm
Quantity 20 Pk.
Width (Metric) 158 mm