missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dehydroepiandrosterone-16,16-d2, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Dehydroepiandrosterone 16,16-d2 |
| CAS | 67034-83-7 |
| Molecular Formula | C19 D2 H26 O2 |
| Quantity | 0.1 g |
| Synonym | Dehydro Epiandrosterone D2 (16,16-D2), Androst-5-en-17-one-16,16-d2, 3-hydroxy-, (3β)-, (3β)-3-Hydroxyandrost-5-en-17-one-16,16-d2, delta;5-Androstene-3β-ol-17-one-16,16-d2, Dehydroepiandrosterone-16,16-D2, DHEA-d2, Dehydroepiandrosterone-d2, Prasterone-d2, Prasterone-16,16-d2 |
| InChI Formula | InChI=1 S/C19H28O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h3,13-16,20 H,4-11H2,1-2H3/t13-,14-,15-,16-,18-,19-/m0/s1/i6D2 |
| SMILES | [2 H]C1([2 H])C[C@H]2 [C@@H]3 CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3 CC[C@]2(C)C1=O |
| IUPAC Name | (3 S,8 R,9 S,10 R,13 S,14 S)-16,16-dideuterio-3-hydroxy-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15-decahydro-1 H-cyclopenta[a]phenanthren-17-one |
| Molecular Weight (g/mol) | 290.4367 |
| Formula Weight | 290.2215 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction