Learn More
Dehydro Nifedipine-d6, TRC
Specifications
Specifications
| Chemical Name or Material | Dehydro Nifedipine-d6 |
| CAS | 125464-52-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C17H10D6N2O6 |
| Quantity | 1 mg |
| Synonym | 3,5-Pyridinedicarboxylic acid','2,6-dimethyl-4-(2-nitrophenyl)-','di(methyl-d3) ester (9CI), Dehydronifedipine-d6, Di(methyl-d3) 2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylate, Dimethyl 2,6-dimethyl-4-(2-nitrophenyl)pyridine-3,5-dicarboxylate D6 (di(methyl D3) ester), Nitrophenylpyridine Analogue of Nifedipine-d6, Dehydro Nifedipine-d6, Oxidized Nifedipine-d6 |
| InChI Formula | InChI=1S/C17H16N2O6/c1-9-13(16(20)24-3)15(14(10(2)18-9)17(21)25-4)11-7-5-6-8-12(11)19(22)23/h5-8H,1-4H3/i3D3,4D3 |
| SMILES | [2H]C([2H])([2H])OC(=O)c1c(C)nc(C)c(C(=O)OC([2H])([2H])[2H])c1c2ccccc2[N+](=O)[O-] |
| Molecular Weight (g/mol) | 350.36 |
| Percent Purity | >95 |