Learn More
Deguelin, TRC
Product Code. 17989534
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17989534 10 mg 10mg
30134927 5 mg 5mg
17919744 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17989534 Supplier Toronto Research Chemicals Supplier No. TRCD22920510MG
 Restricted Item
Estimated Shipment: 03-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 522-17-8
Molecular Formula C23 H22 O6
Molecular Weight (g/mol) 394.42
Synonym (7aS,13aS)-13,13a-Dihydro-9,10-dimethoxy-3,3-dimethyl-3H-bis[1]benzopyrano[3,4-b:6',5'-e]pyran-7(7aH)-one, (-)-Deguelin, (-)-cis-Deguelin;
SMILES COc1cc2OC[C@H]3Oc4c5C=CC(C)(C)Oc5ccc4C(=O)[C@H]3c2cc1OC
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C23H22O6/c1-23(2)8-7-12-15(29-23)6-5-13-21(24)20-14-9-17(25-3)18(26-4)10-16(14)27-11-19(20)28-22(12)13/h5-10,19-20H,11H2,1-4H3/t19-,20+/m1/s1
Formula Weight 394.1416
Percent Purity >95%
Chemical Name or Material Deguelin
Grade HPLC