Learn More
OHAUS™ Defender™ 5000 Washdown - D52 Low Profile Bench Scale, Models TD52XW QX
Product Code. 15948615 Shop All Ohaus Products
Change view
Click to view available options
Model:
D52XW150WQDX5-EU
D52XW150WQDX5-M
D52XW150WQX5-EU
D52XW150WQX5-M
D52XW60WQDX5-EU
D52XW60WQDX5-M
D52XW60WQX5-EU
D52XW60WQX5-M
Unit Size:
1 unit
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
15948615 D52XW150WQX5-EU 1 unit
15918645 D52XW60WQX5-M 1 unit
15928545 D52XW150WQDX5-EU 1 unit
15928645 D52XW150WQX5-M 1 unit
15938615 D52XW60WQX5-EU 1 unit
15958485 D52XW60WQDX5-EU 1 unit
15978545 D52XW60WQDX5-M 1 unit
15988545 D52XW150WQDX5-M 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15948615 Supplier OHAUS™ Supplier No. 30471129
Only null left
Add to basket
Add to basket
OHAUS™ Defender™ 5000 Series multifunctional bench scale is ideal for a multitude of applications. Built durably and equipped with advanced features such as maximum configurability print output, GMP/GLP data output, library/user management and multiple connectivity options.
  • User-friendly model with an easy-to-use keypad, large backlit display and plain text on-screen prompts which guide you through operation.
  • Durable construction with a stainless steel platform, frame, IP67 load cell and IP68 indicator.
  • Defender meets NTEP/Measurement Canada or OIML/EC Type approvals (depending on region).
  • Features common industrial modes and supports a 50,000-item library with 1,000 user profiles for easy data management.
  • Incudes GMP/GLP with time/date for result traceability.
TRUSTED_SUSTAINABILITY
Specifications
Battery Life 20 Hours with Rechargeable Battery
Connection Type Easy access communication ports including RS232 and Micro SD slot (included), second RS232/RS485/USD device, Ethernet and WiFi/Bluetooth, Discrete I/O and Analog Output (sold separately as accessories); GLP/GMP data output with real-time clock
Construction 304-grade stainless steel platform and frame, stainless steel IP68 indicator, IP67 load cell, non-slip adjustable rubber feet
Capacity 150.00 kg
Display Type Backlit LCD
Readability 10.000000 g
Model D52XW150WQX5-EU
Protection Class Stainless Seel IP68 Indicator, IP67 Load Cell
Temperature (Metric) Operating -10°C to +40°C
Weighing Modes Custom, Gram, Kilogram, Metric Tonnes, Ounce, Pound, Pound:Ounce, Tonne
Dimensions (L x W x H) 772 x 500 x 122 mm
Weight (Metric) 20.740 kg