Learn More
OHAUS™ Defender™ 5000 - D52 Bench Scale, Models TD52XW RTV
Product Code. 15908515 Shop All Ohaus Products
Change view
Click to view available options
Model:
D52XW150RTDV3-EU
D52XW150RTDV3-M
D52XW300RTDV3-EU
D52XW300RTDV3-M
D52XW600RTDV3-EU
D52XW600RTDV3-M
Unit Size:
1 unit
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
15908515 D52XW600RTDV3-EU 1 unit
15948595 D52XW150RTDV3-M 1 unit
15958595 D52XW300RTDV3-M 1 unit
15968595 D52XW600RTDV3-M 1 unit
15988505 D52XW150RTDV3-EU 1 unit
15998505 D52XW300RTDV3-EU 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15908515 Supplier OHAUS™ Supplier No. 30444816
Add to basket
Add to basket
OHAUS™ Defender™ 5000 Series multifunctional bench scale is ideal for a multitude of applications. Built durably and equipped with advanced features such as maximum configurability print output, GMP/GLP data output, library/user management and multiple connectivity options.
  • User-friendly model with an easy-to-use keypad, large backlit display and plain text on-screen prompts which guide you through operation.
  • Durable construction with a stainless steel platform, sand gray powder-coated steel frame and aluminum load cell.
  • Defender meets NTEP/Measurement Canada or OIML/EC type approvals (depending on region).
  • Features common industrial modes and supports a 50000-item library with 1000 user profiles for easy data management.
  • Includes GMP/GLP with time/date for result traceability.
TRUSTED_SUSTAINABILITY
Specifications
Battery Life 20 Hours with Rechargeable Battery
Connection Type Easy access communication ports including RS232 and Micro SD slot (included), second RS232/RS485/USD device, Ethernet and WiFi/Bluetooth, Discrete I/O and Analog Output (sold separately as accessories); GLP/GMP data output with real-time clock
Construction Stainless steel (TD52XW) indicator, 304-grade stainless steel platform with sand gray powder-coated steel frame, aluminum load cell, non-slip adjustable rubber feet
Capacity 300.00 kg, 600.00 kg
Display Type Backlit LCD
Readability 20.000000 g, 50.000000 g
Model D52XW600RTDV3-EU
Temperature (Metric) Operating -10°C to +40°C
Weighing Modes Custom, Gram, Kilogram, Metric Tonnes, Ounce, Pound, Pound:Ounce, Tonne
Dimensions (L x W x H) 963 x 600 x 1234 mm
Weight (Metric) 48.000 kg