Learn More
OHAUS™ Defender™ 5000 - D52 Bench Scale, Models TD52XW RQL
Product Code. 15988585 Shop All Ohaus Products
Change view
Click to view available options
Model:
D52XW15RQDL2-EU
D52XW15RQDL2-M
D52XW30RQDL2-EU
D52XW30RQDL2-M
D52XW60RQDL2-EU
D52XW60RQDL2-M
Unit Size:
1 unit
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
15988585 D52XW60RQDL2-M 1 unit
15908505 D52XW15RQDL2-EU 1 unit
15918505 D52XW30RQDL2-EU 1 unit
15928505 D52XW60RQDL2-EU 1 unit
15968585 D52XW15RQDL2-M 1 unit
15978585 D52XW30RQDL2-M 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15988585 Supplier OHAUS™ Supplier No. 30461583
Only null left
Add to basket
Add to basket
OHAUS™ Defender™ 5000 Series multifunctional bench scale is ideal for a multitude of applications. Built durably and equipped with advanced features such as maximum configurability print output, GMP/GLP data output, library/user management and multiple connectivity options.
  • User-friendly model with an easy-to-use keypad, large backlit display and plain text on-screen prompts which guide you through operation.
  • Durable construction with a stainless steel platform, sand gray powder-coated steel frame and aluminum load cell.
  • Defender meets NTEP/Measurement Canada or OIML/EC type approvals (depending on region).
  • Features common industrial modes and supports a 50000-item library with 1000 user profiles for easy data management.
  • Includes GMP/GLP with time/date for result traceability.
TRUSTED_SUSTAINABILITY
Specifications
Battery Life 20 Hours with Rechargeable Battery
Connection Type Easy access communication ports including RS232 and Micro SD slot (included), second RS232/RS485/USD device, Ethernet and WiFi/Bluetooth, Discrete I/O and Analog Output (sold separately as accessories); GLP/GMP data output with real-time clock
Construction Stainless steel (TD52XW) indicator, 304-grade stainless steel platform with sand gray powder-coated steel frame, aluminum load cell, non-slip adjustable rubber feet
Capacity 30.00 kg, 60.00 kg
Display Type Backlit LCD
Readability 10.000000 g, 20.000000 g
Model D52XW60RQDL2-M
Temperature (Metric) Operating -10°C to +40°C
Weighing Modes Gram, Kilogram, Metric Tonnes
Dimensions (L x W x H) 570 x 400 x 934 mm
Weight (Metric) 18.200 kg