Learn More
Ohaus™ Defender™ 3000 Hybrid
Product Code. 15374224 Shop All Ohaus Products
Click to view available options
:
Model: D32XW150BX-EU
Product Code. 15374224
Brand: Ohaus™ 83997633
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
OHAUS™ Defender™ 3000 Hybrid is ideal for general weighing and simple counting applications. Features a simple, yet rugged stainless steel design and 4X/IP66 stainless steel indicator with tactile keys, backlit LCD display, and built-in rechargeable battery.
  • The IP66 stainless steel indicator is built to endure tough industrial elements.
  • The 58-hour rechargeable battery offers continuous operation for multi-day operations.
  • This tough industrial scale features a 304 stainless steel platform, stainless steel column and indicator bracket, painted tubular frame, and aluminum IP67 load cell.
  • Easy-to-read display and basic counting mode allow for simple operation for everyday basic industrial applications.
  • Offered with OIML/EC Type approvals.
TRUSTED_SUSTAINABILITY
Specifications
Battery Life 58 Hours with Rechargeable Battery
Connection Type Easy access communication port with RS232 interface (included)
Construction IP65 stainless steel hinged front cover indicator, stainless steel column and wall bracket, 304 grade stainless steel platform with painted steel tubular frame, IP67 aluminum load cell
Capacity 150.00 kg
Display Type Backlit LCD
Readability 20 g
Model D32XW150BX-EU
Protection Class IP65, NEMA 4X
Temperature (Metric) Operating -10°C to +40°C
Weighing Modes Gram, Kilogram, Ounce, Pound, Pound:Ounce
Dimensions (L x W x H) 740 x 500 x 920 mm
Weight (Metric) 28 kg