Learn More
Metro™ Deep Ledge Utility Carts
Product Code. 11830410
Change view
Click to view available options
Dimensions (L x W x H):
831 x 546 x 1041 mm
984 x 685 x 1041 mm
No. of Shelves:
2
3
Color:
Black
Blue
Gray
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) No. of Shelves Color Unit Size
11830410 831 x 546 x 1041 mm 2 Black Each
11726658 831 x 546 x 1041 mm 3 Gray Each
11776668 984 x 685 x 1041 mm 3 Gray Each
11716658 984 x 685 x 1041 mm 2 Black Each
11756658 831 x 546 x 1041 mm 3 Black Each
11796668 984 x 685 x 1041 mm 3 Blue Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11830410 Supplier Metro™ Supplier No. BC20302DBL
  • ONLINE EXCLUSIVE PRICE
    405.42€
    184.32€ / Pack of 1
    Save 221.10€
    54% Off
Estimated Shipment: 30-12-2026
Useful for a variety of transportation and temporary storage tasks. Metro™ Deep Ledge Utility Carts features a deep ledge design that keeps items from sliding off of the cart.
  • Ultimate cleanability - smooth surfaces are designed to resist staining and wipe clean easily
  • Corrosion-proof shelves - all-polymer shelf will never corrode
  • Contains spills - shelves feature a deep 70mm ledge to contain product or spills
  • Available in four colors - gray, blue, black, or with Microban™ product protection in slate blue
  • Attractive aesthetics allow for use in a variety of applications
  • Sturdy construction - each shelf can hold up to 68kg; each unit can hold up up 181kg
  • Easy to adjust - optional center shelf adjusts in 25mm increments
  • Ergonomic handle, designed for easy use
  • Easy, no-tool assembly
  • Available in 2-shelf and 3-shelf models
  • Microban protection inhibits the growth of bacteria, mold, mildew, and fungi that cause odors, stains and product degradation
  • Shelves are manufactured from injection-molded polyurethane with chrome-plated posts
  • Carts roll on four 102mm diameter thermoplastic, mark-resistant swivel/stem casters
Specifications
Capacity (Metric) Shelf 68 kg
Product Type Laboratory/Utility Cart
Certifications/Compliance NSF
Color Black
Dimensions (L x W x H) 831 x 546 x 1041 mm
No. of Casters 4
No. of Shelves 2