Learn More
GA International Deep-Freeze Labels for Thermal-Transfer Barcode Printers on Large Roll
Product Code. 17811288
Encompass_Preferred
Change view
Click to view available options
Dimensions (LxW):
22 x 15 mm
25.4 x 12.7 mm
31.8 x 22.2 mm
38.1 x 19.1 mm
76.2 x 25.4 mm
Unit Size:
Pack of 1000
Pack of 2000
Pack of 4000
Pack of 5000
Color:
Blue
Brown
Green
Lavender
Lemon
Mint
Orange
Pink
Red
Silver
Yellow
Product Code. Dimensions (LxW) unitSize Color
17811288 38.1 x 19.1 mm Pack of 2000 -
17861188 22 x 15 mm Pack of 4000 Yellow
17819047 76.2 x 25.4 mm Pack of 1000 Blue
17860608 76.2 x 25.4 mm Pack of 1000 Brown
17868857 76.2 x 25.4 mm Pack of 1000 Green
17898957 76.2 x 25.4 mm Pack of 1000 Lavender
17809547 76.2 x 25.4 mm Pack of 1000 Orange
17849157 76.2 x 25.4 mm Pack of 1000 Pink
17860388 76.2 x 25.4 mm Pack of 1000 Red
17850758 76.2 x 25.4 mm Pack of 1000 Silver
17840758 76.2 x 25.4 mm Pack of 1000 Yellow
17869327 25.4 x 12.7 mm Pack of 5000 Blue
17880588 25.4 x 12.7 mm Pack of 5000 Green
17809727 25.4 x 12.7 mm Pack of 5000 Lemon
17831178 25.4 x 12.7 mm Pack of 5000 Pink
17840698 25.4 x 12.7 mm Pack of 5000 Red
17840988 25.4 x 12.7 mm Pack of 5000 Silver
17859527 25.4 x 12.7 mm Pack of 5000 Yellow
17800608 31.8 x 22.2 mm Pack of 1000 Green
17889617 31.8 x 22.2 mm Pack of 1000 Mint
17820898 31.8 x 22.2 mm Pack of 1000 Orange
17860038 31.8 x 22.2 mm Pack of 1000 Red
17849457 31.8 x 22.2 mm Pack of 1000 Yellow
23 options
Product Code. 17811288
Brand: GA International FJT10C32OR
Only null left
Add to basket
Add to basket
Permanent deep-freeze labels for storage of samples in lab refrigerators, ultra-low temperature freezers, dry ice transport and general lab use.
  • Ideal for long-term storage in deep-freeze conditions (-80°C)
  • No curing time, can be stored in freezers immediately after application
  • No additional lamination necessary
  • Labels and printout resist spraying with alcohols and other chemicals
  • Printable with all major thermal-transfer printer brands
  • FDA Compliance: FDA 21 CFR 175.105 for indirect food contact
  • RoHS compliant
  • Waterproof and tearproof
  • Adhere to moist and wet surfaces
  • Resist spraying with alcohols and withstand multiple freeze-thaw cycles
Specifications
Finish Matte
Format Roll
Material Thermoplastic Film
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
Labels per Row 1 Label
Quantity 2000/Pk.
Shape Rectangle
Temperature Range (English) -112°F to +230°F
Temperature Range (Metric) -80°C to +110°C
Dimensions (LxW) 38.1 x 19.1 mm
Length (English) 0.75 in.
Length (Metric) 19.1 mm
Width (English) 1.5 in.
Width (Metric) 38.1 mm
Diameter (English) 3 in.
Diameter (Metric) 76.2 mm