Learn More
Buerkle™ Deep-Form Photographic Tray without Bottom Ribs
Product Code. 10018601 Shop All Bürkle Products
Click to view available options
Dimensions (L x W x H):
260 x 340 x 85 mm
330 x 390 x 100 mm
430 x 530 x 105 mm
520 x 610 x 110 mm
630 x 730 x 110 mm
Color:
Black
Red
White
Unit Size:
Each
Product Code. 10018601
Brand: Buerkle™ 42071050
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use these trays for universal laboratory or storage purposes. Buerkle™ Deep-Form Photographic Tray without Bottom Ribs are made of high quality PVC and available in shallow or deep models in 3 attractive, high-gloss colors: white, red and black.
  • PVC
  • Thick sides, shatterproof
  • Largely resistant to acids and alkalis
  • Liquid can be poured out easily from the tray edges
  • Not suitable for temperature controlled cupboards or dishwashers
  • Temperature resistant from -30 to +60°C
TRUSTED_SUSTAINABILITY
Specifications
Dimensions (L x W x H) 630 x 730 x 110 mm
Color White
Height (Metric) 110 mm
Length (Metric) 510 mm (Inner Base), 630 mm (Outer Base)
Material PVC
Type Photographic Tray
Width (Metric) 610 mm (Inner Base), 730 mm (Outer Base)
Height (English) 4.33 in.
Shape Rectangular