missing translation for 'onlineSavingsMsg'
Learn More

Decyl Beta-D-Glucopyranoside, TRC

Product Code. 30381572
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381572 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381572 Supplier Toronto Research Chemicals Supplier No. TRCD525518500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Decyl Beta-D-Glucopyranoside
CAS 58846-77-8
Molecular Formula C16 H32 O6
Quantity 500 mg
Synonym 1-O-Decyl-β-D-glucopyranoside, AG 10, AG 10 (carbohydrate), APG 10, Decyl Glucoside, Decyl β-D-glucoside, Decyl β-glucopyranoside, Decyl β-glucoside, Oramix NS 10, Plantacare 818, Plantaren PS 100, Plantaren W 2000, n-Decyl β-D-glucopyranoside, n-Decyl-β-glucoside
InChI Formula InChI=1S/C16H32O6/c1-2-3-4-5-6-7-8-9-10-21-16-15(20)14(19)13(18)12(11-17)22-16/h12-20H,2-11H2,1H3/t12-,13-,14+,15-,16-/m1/s1
SMILES CCCCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
IUPAC Name (2R,3R,4S,5S,6R)-2-decoxy-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol) 320.422
Formula Weight 320.22
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.