Learn More
Abcam Decoyinine (Augustmycin A), Nucleoside analog
Product Code. 32118038 Shop All Abcam Products
Change view
Click to view available options
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
32118038 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32118038 Supplier Abcam Supplier No. ab1442381mg
 Restricted Item
Estimated Shipment: 03-12-2026
MW 279.25 Da, Purity >98%. Nucleoside analog. Selective, reversible non-competitive GMP synthetase inhibitor. Reduces intracellular GDP and GTP levels. Induces sporulation. Shows antitumor effects.
MW 279.25 Da, Purity >98%. Nucleoside analog. Selective, reversible non-competitive GMP synthetase inhibitor. Reduces intracellular GDP and GTP levels. Induces sporulation. Shows antitumor effects.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 1 mg