Learn More
Decon Laboratories™ Decomatic™ Cleaning Agent
Product Code. 11731178
Change view
Click to view available options
pH:
13
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Unit Size
11731178 13 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11731178 Supplier Decon Laboratories™ Supplier No. DEC5
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Phosphate-free surface active cleaning agent. Decon Laboratories Decomatic™ Cleaning Agent can be use with laboratory glassware washing machines or for medical and specialized industrial applications.
  • Free from phosphates, enzymes, EDTA or chlorine
  • Biodegradable
  • Bactericidal
  • Non-flammable
  • Non-foaming
  • Totally rinsable
  • Suitable for laboratory glassware, ceramics, plastic (except polycarbonate), rubber, Stainless Steel and iron metals
  • Amphoteric surfactants and stabilizers
Specifications
pH 13
Appearance Pale Amber
Boiling Point 100.8°C
Composition A blend of amphoteric surface active agents, non-phosphate detergent builders, alkalis and sequester
Container Type Polyethylene containers
compliance-icons
Product Identifier
  • DECON DECOMATIC
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: sodium hydroxide