Learn More
Corning™ DeckWorks™ Reload Systems Starter Kits
Product Code. 11393655 Shop All Corning Products
Click to view available options
Volume (Metric):
0.1 to 10 μL
1 to 200 μL
1 to 300 μL
100 to 1000 μL
Color:
Natural
Yellow
Unit Size:
1 set
Product Code. 11393655
Brand: Corning™ 4134
Only null left
Add to basket
Add to basket
Contains one DeckWorks Reload System with standard tips and empty hinged racks
DeckWorks starter kits contain one DeckWorks reload system with standard tips and empty hinged racks.
  • 10 empty racks accompany 10 μL, 200 μL, and 300 μL volume tips.
  • 8 empty racks accompany 1000 μL volume tips.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 100 to 1000 μL
Sterility Non-sterile
Color Natural
For Use With (Application) Designed to allow to maximize storage space and help reduce the production and waste of plastic materials
Format Refill
Tip Style Round
Packaging Starter kit
Graduated Yes
Product Line DeckWorks