Learn More
Dechloro Haloperidol, TRC
Product Code. 30137570
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137570 250 mg 250mg
30008054 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30137570 Supplier Toronto Research Chemicals Supplier No. TRCD226500250MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3109-12-4
Molecular Formula C21H24FNO2
Molecular Weight (g/mol) 341.42
Synonym 1-Butanone, 1-(4-fluorophenyl)-4-(4-hydroxy-4-phenyl-1-piperidinyl)-, Butyrophenone, 4'-fluoro-4-(4-hydroxy-4-phenylpiperidino)- (6CI,7CI,8CI), 1-(4-Fluorophenyl)-4-(4-hydroxy-4-phenyl-1-piperidinyl)-1-butanone, Dechlorohaloperidol, R 1838, 1-(4-Fluorophenyl)-4-(4-hydroxy-4-phenylpiperidin-1-yl)butan-1-one
IUPAC Name 1-(4-fluorophenyl)-4-(4-hydroxy-4-phenylpiperidin-1-yl)butan-1-one
SMILES OC1(CCN(CCCC(=O)c2ccc(F)cc2)CC1)c3ccccc3
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C21H24FNO2/c22-19-10-8-17(9-11-19)20(24)7-4-14-23-15-12-21(25,13-16-23)18-5-2-1-3-6-18/h1-3,5-6,8-11,25H,4,7,12-16H2
Formula Weight 341.1791
Chemical Name or Material Dechloro Haloperidol (Haloperidol Impurity B)