Learn More
Fisherbrand™ Decapping Heads for Pneumatic Hand-held Crimping Tool
Product Code. 11854890 Shop All Fisherbrand Products
Click to view available options
Size:
11 mm
13 mm
20 mm
32 mm
Unit Size:
Each
Product Code. 11854890
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For use with Pneumatic Hand-held Crimping Tool
  • Decapping Head
  • Pneumatic
  • Steel
TRUSTED_SUSTAINABILITY
Specifications
Description 11mm Decapping Head
Type Decapping Head
For Use With (Equipment) Aluminum Cap
Size 11 mm