Learn More
Afora™ Decanting Funnel with Cane
Product Code. p-6983038
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
Diameter (Metric)Top:
80 mm
98 mm
122 mm
152 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Diameter (Metric)Top unitSize
11921099 1000 mL 122 mm Each
11926428 250 mL 80 mm Each
11971079 500 mL 98 mm Each
11931109 2000 mL 152 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11921099 Supplier Afora™ Supplier No. 1098/TFE
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Separates immiscible, heterogenous liquids. Afora™ Decanting Funnel with Cane includes a plastic stopper and Rotaflo TF wrench.
  • Pear-shaped body
  • Cone-shaped bottom
  • Glass with plastic stopcock
  • Includes Rotaflo TF wrench
Specifications
Diameter (Metric)Top 122 mm
Capacity (Metric) 1000 mL
Material Glass
Graduated No