Learn More
Afora™ Decanting Funnel with Cane
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
Diameter (Metric)Top:
80 mm
98 mm
122 mm
152 mm
Unit Size:
Each
Description
- Pear-shaped body
- Cone-shaped bottom
- Glass with plastic stopcock
- Includes Rotaflo TF wrench
Specifications
Specifications
| Diameter (Metric)Top | 152 mm |
| Capacity (Metric) | 2000 mL |
| Material | Glass |
| Graduated | No |