Learn More
Decamethonium Bromide, TRC
Product Code. 30135165
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135165 5 g 5g
30137195 10 g 10g
30010582 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135165 Supplier Toronto Research Chemicals Supplier No. TRCD2124535G
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
This item is not returnable. Consulta la politica di reso
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 541-22-0
Molecular Formula C16H38N2+2
Molecular Weight (g/mol) 258.49
Synonym N,N,N,N',N',N'-Hexamethyl-1,10-decanediaminium, C 10, Decacuran, Decamethonium Bromide, Decamethonium Dibromide, Decamethylene-1,10-bistrimethylammonium Dibromide, Decamethylenebis[trimethylammonium Bromide], Syncurine;
IUPAC Name trimethyl-[10-(trimethylazaniumyl)decyl]azanium
SMILES [Br-].[Br-].C[N+](C)(C)CCCCCCCCCC[N+](C)(C)C
Specifica
Quantity 5 g
InChI Formula InChI=1S/C16H38N2/c1-17(2,3)15-13-11-9-7-8-10-12-14-16-18(4,5)6/h7-16H2,1-6H3/q+2
Formula Weight 416.1402
Chemical Name or Material Decamethonium Bromide
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.