Learn More
Decalin-d18 (cis/trans mixture), CDN
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Chemical Name or Material | Decalin-d18 |
| CAS | 28788-42-3 |
| Molecular Formula | C10 D18 |
| Quantity | 1 g |
| Synonym | Naphthalene-1,2,3,4,5,6,7,8-d8, decahydro-d10- (ACI), Decahydro-d10naphthalene-1,2,3,4,5,6,7,8-d8 (ACI), Naphthalene-d8, decahydro-d10- (8 CI, 9 CI), 1,1,2,2,3,3,4,4,4 A,5,5,6,6,7,7,8,8,8 A-Octadecadeuterionaphthalene, Decahydronaphthalene-d18, Decahydronaphthalene-D18, Decalin-d18 |
| InChI Formula | InChI=1 S/C10H18/c1-2-6-10-8-4-3-7-9(10)5-1/h9-10 H,1-8H2/i1D2,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9 D,10 D |
| SMILES | [2 H]C1([2 H])C([2 H])([2 H])C([2 H])([2 H])C2([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C2([2 H])C1([2 H])[2 H] |
| IUPAC Name | 1,1,2,2,3,3,4,4,4 A,5,5,6,6,7,7,8,8,8 A-octadecadeuterionaphthalene |
| Molecular Weight (g/mol) | 156.3608 |
| Formula Weight | 156.2538 g/mol |
| Show More |