Learn More
De-guanidine Peramivir, TRC
Specifications
Specifications
| Chemical Name or Material | De-guanidine Peramivir |
| CAS | 229614-37-3 |
| Molecular Formula | C14 H26 N2 O4 |
| Quantity | 10 mg |
| Synonym | (1S,2S,3R,4R)-3-[(1S)-1-(Acetylamino)-2-ethylbutyl]-4-amino-2-hydroxycyclopentanecarboxylic Acid |
| InChI Formula | InChI=1S/C14H26N2O4/c1-4-8(5-2)12(16-7(3)17)11-10(15)6-9(13(11)18)14(19)20/h8-13,18H,4-6,15H2,1-3H3,(H,16,17)(H,19,20)/t9-,10+,11+,12-,13+/m0/s1 |
| SMILES | CCC(CC)[C@H](NC(=O)C)[C@H]1[C@H](N)C[C@@H]([C@H]1O)C(=O)O |
| IUPAC Name | (1S,2S,3R,4R)-3-[(1S)-1-acetamido-2-ethylbutyl]-4-amino-2-hydroxycyclopentane-1-carboxylic acid |
| Molecular Weight (g/mol) | 286.367 |
| Formula Weight | 286.189 |