missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ DDT (Human) Recombinant Protein
Human DDT full-length ORF ( AAH05971, 1 a.a. - 118 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001652-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MPFLELDTNLPANRVPAGLEKRLCAAAASILGKPADRVNVTVRPGLAMALSGSTEPCAQLSISSIGVVGTAEDNRSHSAHFFEFLTKELALGQDRILIRFFPLESWQIGKIGTVMTFL
Specifications
AAH05971 | |
D-dopachrome tautomerase | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DDT | |
GST |
1652 | |
Wheat germ expression system | |
10 μg | |
DDCT | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |