missing translation for 'onlineSavingsMsg'
Learn More

DCG IV, TRC

Product Code. 30366228
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30366228 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30366228 Supplier Toronto Research Chemicals Supplier No. TRCD19518010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-[(S)-amino(carboxy)methyl]cyclopropane-1,2-dicarboxylic acid
CAS 147782-19-2
Molecular Formula C7 H9 N O6
Quantity 10 mg
Synonym DCG IV, (2S,2'R,3'R)-2-(2',3'-Dicarboxycyclopropyl)glycine, (1R,2R)-3-[(S)-Aminocarboxymethyl]-1,2-cyclopropanedicarboxylic Acid
InChI Formula InChI=1S/C7H9NO6/c8-4(7(13)14)1-2(5(9)10)3(1)6(11)12/h1-4H,8H2,(H,9,10)(H,11,12)(H,13,14)/t1?,2?,3?,4-/m0/s1
SMILES N[C@@H](C1C(C1C(=O)O)C(=O)O)C(=O)O
IUPAC Name 3-[(S)-amino(carboxy)methyl]cyclopropane-1,2-dicarboxylic acid
Molecular Weight (g/mol) 203.149
Formula Weight 203.043
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.